3,5-Dichloro-2-methylphenylboronic acid
Supplier: Apollo Scientific
Warning
OR11933-1G OR11933-250MG
APOSOR11933-1GEA 328 EUR
APOSOR11933-1G APOSOR11933-250MG
3,5-Dichloro-2-methylphenylboronic acid
3,5-Dichloro-2-methylphenylboronic acid
Formula: C₇H₇BCl₂O₂
Storage Temperature: Ambient
MDL Number: MFCD09800882
CAS Number: 957120-97-7