3-Bromobenzamide

Supplier: Apollo Scientific
Warning

OR11990-25G OR11990-5G
APOSOR11990-25GEA 43.5 EUR
APOSOR11990-25G APOSOR11990-5G
3-Bromobenzamide
3-Bromobenzamide
Formula: C₇H₆BrNO
MW: 200.04 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00017127
CAS Number: 22726-00-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR