2,5-Dibromopyridine
Supplier: Apollo Scientific
Warning
OR12002-100G OR12002-25G
APOSOR12002-100GEA 26.4 EUR
APOSOR12002-100G APOSOR12002-25G
2,5-Dibromopyridine
2,5-Dibromopyridine
Formula: C₅H₃Br₂N
MW: 236.89 g/mol
Boiling Pt: 235 °C
Melting Pt: 92…95 °C
Storage Temperature: Ambient
MDL Number: MFCD00006221
CAS Number: 624-28-2
EINECS: 210-839-6