N-(2-Chloroethyl)-N'-(2,4-dimethylphenyl)urea
Supplier: Apollo Scientific
OR12060-1G OR12060-5G
APOSOR12060-1GEA 137 EUR
APOSOR12060-1G APOSOR12060-5G
N-(2-Chloroethyl)-N'-(2,4-dimethylphenyl)urea
N-(2-Chloroethyl)-N'-(2,4-dimethylphenyl)urea
Formula: C₁₁H₁₅ClN₂O
Storage Temperature: Ambient
MDL Number: MFCD00444204
CAS Number: 162376-66-1