6-(Methylamino)nicotinonitrile
Supplier: Apollo Scientific
Warning
OR12120-1G OR12120-250MG
APOSOR12120-1GEA 48 EUR
APOSOR12120-1G APOSOR12120-250MG
6-(Methylamino)nicotinonitrile
6-(Methylamino)nicotinonitrile
Formula: C₇H₇N₃
MW: 133.15 g/mol
Melting Pt: 131…133 °C
Storage Temperature: Ambient
MDL Number: MFCD08689756
CAS Number: 261715-36-0
UN: 2811
ADR: 6.1 III