8-Methyl-4-quinolinol
Supplier: Apollo Scientific
Warning
Synonyms: 4-Hydroxy-8-methylquinoline, 8-Methylquinolin-4-ol
OR12150-1G OR12150-5G
APOSOR12150-1GEA 27.7 EUR
APOSOR12150-1G APOSOR12150-5G
8-Methyl-4-quinolinol
8-Methyl-4-quinolinol
Formula: C₁₀H₉NO
Melting Pt: 213…215 °C
Storage Temperature: Ambient
MDL Number: MFCD00272344
CAS Number: 23432-44-2