2-(4-Chlorophenoxy)acetohydrazide
Supplier: Apollo Scientific
Warning
OR12181-1G OR12181-5G
APOSOR12181-1GEA 58 EUR
APOSOR12181-1G APOSOR12181-5G
2-(4-Chlorophenoxy)acetohydrazide
2-(4-Chlorophenoxy)acetohydrazide
Formula: C₈H₉ClN₂O₂
Melting Pt: 165…167 °C
Storage Temperature: Ambient
MDL Number: MFCD00705185
CAS Number: 2381-75-1