Morpholino(piperidin-4-yl)methanone hydrochloride 90%
Supplier: Apollo Scientific
Warning
Synonyms: 4-(Piperidin-4-ylcarbonyl)morpholine hydrochloride
OR12322-1G OR12322-250MG
APOSOR12322-1GEA 128 EUR
APOSOR12322-1G APOSOR12322-250MG
Morpholino(piperidin-4-yl)methanone hydrochloride 90%
Morpholino(piperidin-4-yl)methanone hydrochloride
Formula: C₁₀H₁₉ClN₂O₂
MW: 234.7231 g/mol
Melting Pt: 279…286 °C
Storage Temperature: Ambient
MDL Number: MFCD05864747
CAS Number: 94467-73-9