6-Bromoindole-3-carboxaldehyde
Supplier: Apollo Scientific
Warning
OR12354-1G OR12354-250MG
APOSOR12354-1GEA 24.8 EUR
APOSOR12354-1G APOSOR12354-250MG
6-Bromoindole-3-carboxaldehyde
6-Bromoindole-3-carboxaldehyde
Formula: C₉H₆BrNO
Melting Pt: 200…206 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00792689
CAS Number: 17826-04-9