5-[3-(Bromomethyl)phenyl]-3-methyl-1,2,4-oxadiazole 97%
Supplier: Apollo Scientific
Danger
OR12432-1G OR12432-250MG
APOSOR12432-1GEA 429 EUR
APOSOR12432-1G APOSOR12432-250MG
5-[3-(Bromomethyl)phenyl]-3-methyl-1,2,4-oxadiazole 97%
5-[3-(Bromomethyl)phenyl]-3-methyl-1,2,4-oxadiazole
Formula: C₁₀H₉BrN₂O
Melting Pt: 66…67.5 °C
Storage Temperature: Ambient
MDL Number: MFCD09817470
CAS Number: 926921-57-5
UN: 3261
ADR: 8,III