3-Isothiocyanatopyridine 98%
Supplier: Apollo Scientific
Danger
OR1254-1G OR1254-5G OR1254-25G
APOSOR1254-1GEA 63.8 EUR
APOSOR1254-1G APOSOR1254-5G APOSOR1254-25G
3-Isothiocyanatopyridine 98%
3-Isothiocyanatopyridine
Formula: C₆H₄N₂S
Boiling Pt: 231…233 °C
Density: 1.22 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00052351
CAS Number: 17452-27-6
UN: 2927
ADR: 6.1+8,II