Homophthalic anhydride
Supplier: Apollo Scientific
Warning
OR12643-100G OR12643-25G OR12643-5G
APOSOR12643-100GEA 285 EUR
APOSOR12643-100G APOSOR12643-25G APOSOR12643-5G
Homophthalic anhydride
Homophthalic anhydride
Formula: C₉H₆O₃
MW: 162.14 g/mol
Melting Pt: 140…142 °C
Storage Temperature: Ambient
MDL Number: MFCD00006894
CAS Number: 703-59-3
EINECS: 211-873-4