(4-Chlorophenyl)methanesulphonyl chloride
Supplier: Apollo Scientific
Danger
Synonyms: 4-Chloro-α-toluenesulphonyl chloride
OR12681-1G OR12681-25G OR12681-5G
APOSOR12681-1GEA 21.9 EUR
APOSOR12681-1G APOSOR12681-25G APOSOR12681-5G
(4-Chlorophenyl)methanesulphonyl chloride
(4-Chlorophenyl)methanesulphonyl chloride
Formula: C₇H₆Cl₂O₂S
Melting Pt: 93…97 °C
Storage Temperature: Ambient
MDL Number: MFCD01631927
CAS Number: 6966-45-6
UN: 3261
ADR: 8,II