4-Hydroxy-2-quinolinecarboxylic acid hydrate
Supplier: Apollo Scientific
Warning
Synonyms: 4-Hydroxyquinaldic acid hydrate, Kynurenic acid hydrate
OR12784-1G OR12784-5G
APOSOR12784-1GEA 23.2 EUR
APOSOR12784-1G APOSOR12784-5G
4-Hydroxy-2-quinolinecarboxylic acid hydrate
4-Hydroxy-2-quinolinecarboxylic acid hydrate
Formula: C₁₀H₇NO₃·xH₂O
MW: 189.17 g/mol (anhy)
Melting Pt: 275 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00006753
CAS Number: 345909-35-5
Merck Index: 13,05342