1-Bromo-4-ethylbenzene
Supplier: Apollo Scientific
Warning
Synonyms: 4-(Bromophenyl)ethane
OR13057-100G OR13057-25G OR13057-500G
APOSOR13057-100GEA 31.9 EUR
APOSOR13057-100G APOSOR13057-25G APOSOR13057-500G
1-Bromo-4-ethylbenzene
1-Bromo-4-ethylbenzene
Formula: C₈H₉Br
Boiling Pt: 203…204 °C
Density: 1.341 g/cm³ (25 °C)
Flash Pt: 63 °C
Storage Temperature: Ambient
MDL Number: MFCD00000111
CAS Number: 1585-07-5
EINECS: 216-439-8