(5-Bromo-2-pyridinyl)methanol
Supplier: Apollo Scientific
Warning
Synonyms: 5-Bromo-2-(hydroxymethyl)pyridine, 5-Bromo-2-hydroxymethylpyridine
OR13061 OR13061-5G OR13061-1G OR13061-10G
XXXXOR13061EA 0 EUR
XXXXOR13061 APOSOR13061-5G APOSOR13061-1G APOSOR13061-10G
(5-Bromo-2-pyridinyl)methanol
(5-Bromo-2-pyridinyl)methanol
Formula: C₆H₆BrNO
MW: 188.0200 g/mol
Melting Pt: 58…62 °C
Storage Temperature: Ambient
MDL Number: MFCD04035597
CAS Number: 88139-91-7