α,2,4-Trichlorotoluene 98%
Supplier: Apollo Scientific
Danger
OR13170-100G OR13170-500G
APOSOR13170-100GEA 61.5 EUR
APOSOR13170-100G APOSOR13170-500G
α,2,4-Trichlorotoluene 98%
α,2,4-Trichlorotoluene
Formula: C₇H₅Cl₃
Boiling Pt: 248 °C
Melting Pt: –2.6 °C
Density: 1.407 g/cm³ (25 °C)
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00000895
CAS Number: 94-99-5
EINECS: 202-381-0
UN: 3265
ADR: 8,III