N-(2-Aminoethyl)morpholine
Supplier: Apollo Scientific
Danger
Synonyms: 4-(2-Aminoethyl)morpholine, 2-Morpholinoethanamine
OR13189-250G OR13189-50G
APOSOR13189-250GEA 145 EUR
APOSOR13189-250G APOSOR13189-50G
N-(2-Aminoethyl)morpholine
N-(2-Aminoethyl)morpholine
Formula: C₆H₁₄N₂O
MW: 130.19 g/mol
Boiling Pt: 205 °C
Melting Pt: 24…26 °C
Density: 0.992 g/cm³ (25 °C)
Flash Pt: 86 °C
Storage Temperature: Ambient
MDL Number: MFCD00006182
CAS Number: 2038-03-1
EINECS: 218-011-6
UN: 2923
ADR: 8+6.1 II