Ethyl-3-amino-1H-pyrazole-4-carboxylate
Supplier: Apollo Scientific
Warning
Synonyms: 3-Amino-4-carbethoxypyrazole, Ethyl 3-amino-1H-pyrazole-4-carboxylate
OR13363-25G OR13363-5G
APOSOR13363-25GEA 24.8 EUR
APOSOR13363-25G APOSOR13363-5G
Ethyl-3-amino-1H-pyrazole-4-carboxylate
Ethyl 3-amino-1H-pyrazole-4-carboxylate
Formula: C₆H₉N₃O₂
MW: 155.16 g/mol
Melting Pt: 104…107 °C
Storage Temperature: Ambient
MDL Number: MFCD00005238
CAS Number: 6994-25-8
EINECS: 230-262-3
UN: 3077
ADR: 9+EHS III