5-Benzyloxyindole-3-carbaldehyde
Supplier: Apollo Scientific
Warning
Synonyms: 5-(Benzyloxy)-1H-indole-3-carbaldehyde, 5-benzyloxy-1h-indole-3-carbaldehyde, 5-Benzyloxyindole-3-carboxaldehyde
OR1344-1G OR1344-25G OR1344-5G
APOSOR1344-1GEA 21.9 EUR
APOSOR1344-1G APOSOR1344-25G APOSOR1344-5G
5-Benzyloxyindole-3-carbaldehyde
5-Benzyloxyindole-3-carbaldehyde
Formula: C₁₆H₁₃NO₂
Melting Pt: 239…242 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00014562
CAS Number: 6953-22-6
EINECS: 230-134-7