Diethyl malonate 99%
Supplier: Apollo Scientific
Warning
OR1354-10KG OR1354-2.5KG OR1354-500G
APOSOR1354-10KGEA 290 EUR
APOSOR1354-10KG APOSOR1354-2.5KG APOSOR1354-500G
Diethyl malonate 99%
Diethyl malonate
Formula: C₇H₁₂O₄
Boiling Pt: 199 °C
Melting Pt: –51 °C
Density: 1.055 g/cm³ (25 °C)
Flash Pt: 90 °C
Storage Temperature: Ambient
MDL Number: MFCD00009195
CAS Number: 105-53-3
EINECS: 203-305-9