2-Methyl-1-butene-3-yne 97%
Supplier: Apollo Scientific
Danger
OR13594-25G OR13594-5G
APOSOR13594-25GEA 0 EUR
APOSOR13594-25G APOSOR13594-5G
2-Methyl-1-butene-3-yne 97%
2-Methyl-1-butene-3-yne
Formula: C₅H₆
Boiling Pt: 32…33 °C
Melting Pt: –113 °C
Density: 0.694 g/cm³ (25 °C)
Flash Pt: –45 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008599
CAS Number: 78-80-8
EINECS: 201-144-9
UN: 3295
ADR: 3,I