3-Chloro-2-hydroxypyridine
Supplier: Apollo Scientific
Warning
Synonyms: 3-Chloro-2-pyridinol
OR13614-1G OR13614-5G OR13614-25G
APOSOR13614-1GEA 21.9 EUR
APOSOR13614-1G APOSOR13614-5G APOSOR13614-25G
3-Chloro-2-hydroxypyridine
3-Chloro-2-hydroxypyridine
Formula: C₅H₄ClNO
Melting Pt: 177 °C
Storage Temperature: Ambient
MDL Number: MFCD01646109
CAS Number: 13466-35-8