4-Nitropyridine-N-oxide
Supplier: Apollo Scientific
Danger
Synonyms: 4-Nitropyridine N-oxide
OR13621-100G OR13621-25G
APOSOR13621-100GEA 59.6 EUR
APOSOR13621-100G APOSOR13621-25G
4-Nitropyridine-N-oxide
4-Nitropyridine N-oxide
Formula: C₅H₄N₂O₃
MW: 140.1 g/mol
Boiling Pt: 397
Melting Pt: 159…162 °C
Storage Temperature: Ambient
MDL Number: MFCD00006206
CAS Number: 1124-33-0
EINECS: 214-395-4
UN: 2811
ADR: 6.1 III