(1-(tert-Butoxycarbonyl)-1H-pyrazol-4-yl)boronic acid 98%
Supplier: Apollo Scientific
Synonyms: N-Boc-1H-pyrazole-4-boronic acid, 1-tert-Butoxycarbonyl-1H-pyrazole-4-boronic acid, 1H-Pyrazole-4-boronic acid, N1-BOC protected
OR13663-10G OR13663-25G OR13663-5G
APOSOR13663-10GEA 219 EUR
APOSOR13663-10G APOSOR13663-25G APOSOR13663-5G
(1-(tert-Butoxycarbonyl)-1H-pyrazol-4-yl)boronic acid 98%
(1-(tert-Butoxycarbonyl)-1H-pyrazol-4-yl)boronic acid
Formula: C₈H₁₃BN₂O₄
MW: 212.01 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09951915
CAS Number: 1188405-87-9