Benzoxazole
Supplier: Apollo Scientific
Warning
Synonyms: 1,3-Benzoxazole
OR13697-25G OR13697-100G OR13697-500G
APOSOR13697-25GEA 21.9 EUR
APOSOR13697-25G APOSOR13697-100G APOSOR13697-500G
Benzoxazole
Benzoxazole
Formula: C₇H₅NO
MW: 119.12 g/mol
Boiling Pt: 181…183 °C
Melting Pt: 27…31 °C
Density: 1.175 g/cm³ (25 °C)
Flash Pt: 58 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005765
CAS Number: 273-53-0
EINECS: 205-988-9
UN: 1325
ADR: 4.1 III