2,4-Dihydroxy-3-nitropyridine
Supplier: Apollo Scientific
Warning
OR13715-100G OR13715-10G OR13715-25G
APOSOR13715-100GEA 63.8 EUR
APOSOR13715-100G APOSOR13715-10G APOSOR13715-25G
2,4-Dihydroxy-3-nitropyridine
2,4-Dihydroxy-3-nitropyridine
Formula: C₅H₄N₂O₄
Melting Pt: 265 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD01075671
CAS Number: 89282-12-2