1,1,2-Trimethyl-1H-benz[e]indole 99%
Supplier: Apollo Scientific
Warning
Synonyms: 1,1,2-Trimethyl-1H-benzo[e]indole
OR13722-5G OR13722-100G OR13722-25G
APOSOR13722-5GEA 21.9 EUR
APOSOR13722-5G APOSOR13722-100G APOSOR13722-25G
1,1,2-Trimethyl-1H-benz[e]indole 99%
1,1,2-Trimethyl-1H-benz[e]indole
Formula: C₁₅H₁₅N
MW: 209.29 g/mol
Melting Pt: 111…117 °C
Density: 0.7 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00082627
CAS Number: 41532-84-7