3-(4-Morpholinyl)aniline 97+%
Supplier: Apollo Scientific
Warning
OR1383-1G OR1383-250MG OR1383-5G
APOSOR1383-1GEA 29 EUR
APOSOR1383-1G APOSOR1383-250MG APOSOR1383-5G
3-(4-Morpholinyl)aniline 97+%
3-(4-Morpholinyl)aniline
Formula: C10H14N2O
MW: 178.2300 g/mol
Melting Pt: 117…121 °C
Storage Temperature: Ambient
MDL Number: MFCD03197165
CAS Number: 159724-40-0
UN: 2811
ADR: 6.1 III