Dimethylamine 40% in aqueous solution
Supplier: Apollo Scientific
Danger
OR13973-100ML OR13973-500ML
APOSOR13973-100MLEA 0 EUR
APOSOR13973-100ML APOSOR13973-500ML
Dimethylamine 40% in aqueous solution
Dimethylamine
Formula: C₂H₇N
Boiling Pt: 52 °C
Melting Pt: –37 °C
Density: 0.89 g/cm³ (25 °C)
Flash Pt: 16 °C
Storage Temperature: Ambient
MDL Number: MFCD00008288
CAS Number: 124-40-3
EINECS: 204-697-4
UN: 1160
ADR: 3+8,II