Ethyl-4-{2-hydroxy-2-[4-(methylsulphonyl)phenyl]ethyl}tetrahydro-1(2H)-pyrazinecarboxylate
Supplier: Apollo Scientific
Synonyms: Ethyl 4-{2-hydroxy-2-[4-(methylsulphonyl)phenyl]ethyl}piperazine-1-carboxylate
OR14122-10G OR14122-1G OR14122-5G
APOSOR14122-10GEA 1350 EUR
APOSOR14122-10G APOSOR14122-1G APOSOR14122-5G
Ethyl-4-{2-hydroxy-2-[4-(methylsulphonyl)phenyl]ethyl}tetrahydro-1(2H)-pyrazinecarboxylate
Ethyl-4-{2-hydroxy-2-[4-(methylsulphonyl)phenyl]ethyl}tetrahydro-1(2H)-pyrazinecarboxylate
Formula: C₁₆H₂₄N₂O₅S
MW: 356.44 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08689707
CAS Number: 924869-23-8