Thiazole-2-carbaldehyde 95+%
Supplier: Apollo Scientific
Warning
Synonyms: 2-Thiazolecarboxaldehyde, Thiazole-2-carboxaldehyde
OR14343-1G OR14343-5G OR14343-250MG
APOSOR14343-1GEA 24.8 EUR
APOSOR14343-1G APOSOR14343-5G APOSOR14343-250MG
Thiazole-2-carbaldehyde 95+%
Thiazole-2-carbaldehyde
Formula: C₄H₃NOS
MW: 113.13 g/mol
Boiling Pt: 61…63 °C/15mmHg
Density: 1.288 g/cm³ (25 °C)
Flash Pt: 68 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00142924
CAS Number: 10200-59-6