2-Bromo-4-chloroanisole
Supplier: Apollo Scientific
Warning
Synonyms: 2-Bromo-4-chloro-1-methoxybenzene
OR1469-100G OR1469-250G OR1469-500G
APOSOR1469-100GEA 50.9 EUR
APOSOR1469-100G APOSOR1469-250G APOSOR1469-500G
2-Bromo-4-chloroanisole
2-Bromo-4-chloroanisole
Formula: C₇H₆BrClO
MW: 221.48 g/mol
Boiling Pt: 127 °C/17.2mmHg
Density: 1.63 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00079705
CAS Number: 60633-25-2