1-Bromonaphthalene 98%
Supplier: Apollo Scientific
Warning
Synonyms: α-Bromonaphthalene
OR14767-1KG OR14767-250G
APOSOR14767-1KGEA 134 EUR
APOSOR14767-1KG APOSOR14767-250G
1-Bromonaphthalene 98%
1-Bromonaphthalene
Formula: C₁₀H₇Br
Boiling Pt: 279…281 °C
Melting Pt: –2 to 0 °C
Density: 1.48 g/cm³ (25 °C)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00003868
CAS Number: 90-11-9
EINECS: 201-965-2