Methyl-6-quinolyl ether 97%
Supplier: Apollo Scientific
Warning
OR14908-25G OR14908-5G
APOSOR14908-25GEA 75.4 EUR
APOSOR14908-25G APOSOR14908-5G
Methyl-6-quinolyl ether 97%
Methyl 6-quinolyl ether
Formula: C₁₀H₉NO
Boiling Pt: 140…146 °C/15mmHg
Melting Pt: 17…20 °C
Density: 1.152 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00006800
CAS Number: 5263-87-6
EINECS: 226-077-2