(3-Chloro-4-methoxyphenyl)methanamine hydrochloride 98%

Supplier: Apollo Scientific
Danger

OR1501-5G OR1501-100G OR1501-25G
APOSOR1501-5GEA 23 EUR
APOSOR1501-5G APOSOR1501-100G APOSOR1501-25G
(3-Chloro-4-methoxyphenyl)methanamine hydrochloride 98%
(3-Chloro-4-methoxyphenyl)methanamine hydrochloride
Formula: C₈H₁₁Cl₂NO
Melting Pt: 250…255 °C
Storage Temperature: Ambient
MDL Number: MFCD01211460
CAS Number: 41965-95-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR