2-(1H-Pyrazol-3-yl)phenol 96%
Supplier: Apollo Scientific
Warning
Synonyms: 3-(2-Hydroxyphenyl)pyrazole
OR15021-1G OR15021-500MG OR15021-5G
APOSOR15021-1GEA 58 EUR
APOSOR15021-1G APOSOR15021-500MG APOSOR15021-5G
2-(1H-Pyrazol-3-yl)phenol 96%
2-(1H-Pyrazol-3-yl)phenol
Formula: C₉H₈N₂O
MW: 160.18 g/mol
Melting Pt: 90…94 °C
Storage Temperature: Ambient
MDL Number: MFCD03093876
CAS Number: 34810-67-8
EINECS: 252-222-4