Methyl-2-(4-bromophenyl)imidazo[1,2-a]pyridine-8-carboxylate
Supplier: Apollo Scientific
OR15102-1G OR15102-250MG
APOSOR15102-1GEA 433 EUR
APOSOR15102-1G APOSOR15102-250MG
Methyl-2-(4-bromophenyl)imidazo[1,2-a]pyridine-8-carboxylate
Methyl 2-(4-bromophenyl)imidazo[1,2-a]pyridine-8-carboxylate
Formula: C₁₅H₁₁BrN₂O₂
Melting Pt: 134…136 °C
Storage Temperature: Ambient
MDL Number: MFCD08444011
CAS Number: 133427-41-5