Thieno[2,3-b]pyridine-2-carboxylic acid
Supplier: Apollo Scientific
Warning
OR15126-1G OR15126-5G OR15126-250MG
APOSOR15126-1GEA 69.6 EUR
APOSOR15126-1G APOSOR15126-5G APOSOR15126-250MG
Thieno[2,3-b]pyridine-2-carboxylic acid
Thieno[2,3-b]pyridine-2-carboxylic acid
Formula: C₈H₅NO₂S
MW: 179.2 g/mol
Melting Pt: 293…294 °C
Storage Temperature: Ambient
MDL Number: MFCD07371381
CAS Number: 59944-76-2