Ethyl 1-(5-chloro-6-oxo-1,6-dihydro-4-pyridazinyl)-4-piperidinecarboxylate
Supplier: Apollo Scientific
OR15260-10G OR15260-1G OR15260-5G
APOSOR15260-10GEA 894 EUR
APOSOR15260-10G APOSOR15260-1G APOSOR15260-5G
Ethyl 1-(5-chloro-6-oxo-1,6-dihydro-4-pyridazinyl)-4-piperidinecarboxylate
Ethyl-1-(5-chloro-6-oxo-1,6-dihydro-4-pyridazinyl)-4-piperidinecarboxylate
Formula: C₁₂H₁₆ClN₃O₃
MW: 285.73 g/mol
Melting Pt: 174…175 °C
Storage Temperature: Ambient
MDL Number: MFCD09864820
CAS Number: 952183-76-5