[1-(4,6-Dimethyl-2-pyrimidinyl)-5-(hydroxymethyl)-1H-1,2,3-triazol-4-yl]methanol
Supplier: Apollo Scientific
OR15402-10G OR15402-1G OR15402-5G
APOSOR15402-10GEA 1350 EUR
APOSOR15402-10G APOSOR15402-1G APOSOR15402-5G
[1-(4,6-Dimethyl-2-pyrimidinyl)-5-(hydroxymethyl)-1H-1,2,3-triazol-4-yl]methanol
[1-(4,6-Dimethyl-2-pyrimidinyl)-5-(hydroxymethyl)-1H-1,2,3-triazol-4-yl]methanol
Formula: C₁₀H₁₃N₅O₂
Melting Pt: 172…174 °C
Storage Temperature: Ambient
MDL Number: MFCD06200948
CAS Number: 886361-66-6