5-Bromo-2-methoxythiazole
Supplier: Apollo Scientific
Warning
OR15561-1G OR15561-5G
APOSOR15561-1GEA 72.5 EUR
APOSOR15561-1G APOSOR15561-5G
5-Bromo-2-methoxythiazole
5-Bromo-2-methoxythiazole
Formula: C₄H₄BrNOS
MW: 194.06 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09025733
CAS Number: 446287-05-4