3-Bromo-5-(chloromethyl)isoxazole 95%
Supplier: Apollo Scientific
Warning
OR15587-1G OR15587-500MG
APOSOR15587-1GEA 340 EUR
APOSOR15587-1G APOSOR15587-500MG
3-Bromo-5-(chloromethyl)isoxazole 95%
3-Bromo-5-(chloromethyl)isoxazole
Formula: C₄H₃BrClNO
Boiling Pt: 110 °C/20mmHg
Storage Temperature: Refrigerator
MDL Number: MFCD06660137
CAS Number: 124498-15-3