O-Benzylhydroxylamine 97%

Supplier: Apollo Scientific
Danger

OR15742-100G OR15742-25G
APOSOR15742-100GEA 162 EUR
APOSOR15742-100G APOSOR15742-25G
O-Benzylhydroxylamine 97%
O-Benzylhydroxylamine
Formula: C₇H₉NO
Boiling Pt: 118…119 °C/30mmHg
Density: 1.058 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00221709
CAS Number: 622-33-3
UN: 2735
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR