Acetylacetone
Supplier: Apollo Scientific
Warning
Synonyms: ACAC, 2,4-Pentanedione
OR16430-2.5L OR16430-500ML
APOSOR16430-2.5LEA 0 EUR
APOSOR16430-2.5L APOSOR16430-500ML
Acetylacetone
Acetylacetone
Formula: CH₃COCH₂COCH₃
Boiling Pt: 140…141 °C
Melting Pt: –23 °C
Density: 0.975 g/cm³ (25 °C)
Flash Pt: 38 °C
Storage Temperature: Ambient
MDL Number: MFCD00008787
CAS Number: 123-54-6
EINECS: 204-634-0
UN: 2310
ADR: 3+6.1,III