4-Bromo-1H-benzoimidazole 98%
Supplier: Apollo Scientific
Warning
OR16442-1G OR16442-5G OR16442-25G
APOSOR16442-1GEA 49.3 EUR
APOSOR16442-1G APOSOR16442-5G APOSOR16442-25G
4-Bromo-1H-benzoimidazole 98%
4-Bromo-1H-benzoimidazole
Formula: C₇H₅BrN₂
MW: 197.0320 g/mol
Melting Pt: 166…168 °C
Storage Temperature: Ambient
MDL Number: MFCD09754164
CAS Number: 83741-35-9