2-Piperidinonicotinic acid
Supplier: Apollo Scientific
Warning
OR16443-1G OR16443-250MG OR16443-500MG
APOSOR16443-1GEA 91.5 EUR
APOSOR16443-1G APOSOR16443-250MG APOSOR16443-500MG
2-Piperidinonicotinic acid
2-Piperidinonicotinic acid
Formula: C₁₁H₁₄N₂O₂
Melting Pt: 102…104 °C
Storage Temperature: Ambient
MDL Number: MFCD02181988
CAS Number: 78253-61-9