3-Aminopentanoic acid
Supplier: Apollo Scientific
Warning
OR16680-1G OR16680-5G OR16680-10G
APOSOR16680-1GEA 89.9 EUR
APOSOR16680-1G APOSOR16680-5G APOSOR16680-10G
3-Aminopentanoic acid
3-Aminopentanoic acid
Formula: C5H11NO2
Storage Temperature: Ambient
CAS Number: 18664-78-3