tert-Butyl-3-(hydroxymethyl)-6-methoxy-1H-indole-1-carboxylate 96%
Supplier: Apollo Scientific
Synonyms: 3-(Hydroxymethyl)-6-methoxy-1H-indole, N-BOC protected
OR1695-1G OR1695-5G
APOSOR1695-1GEA 139 EUR
APOSOR1695-1G APOSOR1695-5G
tert-Butyl-3-(hydroxymethyl)-6-methoxy-1H-indole-1-carboxylate 96%
tert-Butyl-3-(hydroxymethyl)-6-methoxy-1H-indole-1-carboxylate
Formula: C₁₅H₁₉NO₄
Storage Temperature: Ambient
CAS Number: 914349-08-9