3-Aminophenol
Supplier: Apollo Scientific
Danger
Synonyms: 3-Hydroxyaniline, m-Aminophenol
OR16952-1KG OR16952-500G OR16952-100G
APOSOR16952-1KGEA 102 EUR
APOSOR16952-1KG APOSOR16952-500G APOSOR16952-100G
3-Aminophenol
3-Aminophenol
Formula: C₆H₇NO
MW: 109.13 g/mol
Boiling Pt: 164 °C/11mmHg
Melting Pt: 120…124 °C
Flash Pt: 92
Storage Temperature: Ambient
MDL Number: MFCD00007786
CAS Number: 591-27-5
EINECS: 209-711-2
UN: 2512
ADR: 6.1 III
Merck Index: 13,00459